Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mipp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300153.1 | NP_001287082.1 | 5 - 6 | 943 | 151 | |
NM_079385.4 | NP_524109.2 | 5 - 6 | 943 | 151 | ||
NM_168672.3 | NP_730177.1 | 5 - 6 | 884 | 151 | ||
GenBank mRNA | AY071426.1 | - | 5 - 6 | 880 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |