Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104176.2 | NP_001097646.1 | 4 - 5 | 1893 | 71 | |
NM_001275126.1 | NP_001262055.1 | 4 - 5 | 1893 | 71 | ||
NM_079435.4 | NP_524159.1 | 4 - 5 | 1904 | 71 | ||
GenBank mRNA | BT024203.1 | - | 4 - 5 | 1916 | 71 | |
M32660.1 | - | 3 - 4 | 1282 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |