Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CycH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079483.3 | NP_524207.1 | 2 - 3 | 529 | 70 | |
GenBank mRNA | AF074334.1 | - | 3 - 4 | 600 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |