Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CycH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079483.3 | NP_524207.1 | 3 - 4 | 745 | 68 | |
GenBank mRNA | AF074334.1 | - | 4 - 5 | 816 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |