Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prosap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259386.2 | NP_001246315.2 | 10 - 11 | 5365 | 62 | |
NM_001299473.1 | NP_001286402.1 | 10 - 11 | 5122 | 62 | ||
NM_001299474.1 | NP_001286403.1 | 9 - 10 | 4927 | 62 | ||
NM_137081.5 | NP_610925.3 | 11 - 12 | 5560 | 62 | ||
GenBank mRNA | BT044467.1 | - | 11 - 12 | 4643 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |