Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dh44-R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137116.3 | NP_610960.1 | 9 - 10 | 1823 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Drosophila melanogaster |