Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spred | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274050.2 | NP_001260979.1 | 4 - 5 | 856 | 63 | |
NM_137144.4 | NP_610988.1 | 4 - 5 | 856 | 63 | ||
GenBank mRNA | AY069866.1 | - | 4 - 5 | 805 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |