Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
TaqMan
®
Gene Expression Assays
›
Search Tool
›
Search Results
› Dm01824433_g1
See other ZASP52 GE Assays ›
Gene Symbol:
Zasp52
Gene Name:
Z band alternatively spliced PDZ-motif protein 52
Gene Aliases:
CG12968, CG12969, CG30084, CG8242, Dmel\CG30084, Dmel_CG30084, Z(210), ZASP52, Zasp, Zbp210, dzasp, tun, tungus, zasp, zasp52
Chromosome Location:
Chr.2R: 15762565 - 15817004 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0265991
Assay ID
Dm01824433_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Zasp52
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001032249.4
NP_001027420.2
3 - 4
469
72
NM_001032250.3
NP_001027421.1
3 - 4
469
72
NM_001043086.3
NP_001036551.1
3 - 4
469
72
NM_001169692.2
NP_001163163.1
3 - 4
469
72
NM_001274076.1
NP_001261005.1
3 - 4
469
72
NM_001274077.1
NP_001261006.1
3 - 4
469
72
NM_001274078.1
NP_001261007.1
3 - 4
469
72
NM_001274079.1
NP_001261008.1
3 - 4
469
72
NM_001274080.1
NP_001261009.1
3 - 4
469
72
NM_001274082.1
NP_001261011.1
3 - 4
469
72
NM_001274083.1
NP_001261012.1
3 - 4
469
72
NM_001274084.1
NP_001261013.1
3 - 4
469
72
NM_001274085.1
NP_001261014.1
3 - 4
469
72
NM_001274086.1
NP_001261015.1
3 - 4
469
72
NM_001299538.1
NP_001286467.1
3 - 4
469
72
NM_001316421.1
NP_001303350.1
3 - 4
469
72
NM_001316423.1
NP_001303352.1
3 - 4
525
72
NM_001382054.1
NP_001369093.1
3 - 4
469
72
NM_137214.5
NP_611058.4
3 - 4
469
72
NM_145757.3
NP_665700.2
3 - 4
525
72
GenBank mRNA
BT001752.1
-
3 - 4
466
72
BT022171.2
-
3 - 4
449
72
BT081376.1
-
3 - 4
447
72
JN034422.1
-
3 - 4
449
72
JN034423.1
-
3 - 4
330
72
JN034424.1
-
3 - 4
465
72
JN034425.1
-
3 - 4
449
72
JN034426.1
-
3 - 4
456
72
JN034427.1
-
3 - 4
465
72
JN034428.1
-
3 - 4
437
72
JN034429.1
-
3 - 4
456
72
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Drosophila melanogaster
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell-matrix adhesion
actin cytoskeleton organization
myofibril assembly
muscle structure development
actin binding
actinin binding
muscle alpha-actinin binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0