Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169239.2 | NP_001162710.2 | 11 - 12 | 4627 | 66 | |
NM_080341.4 | NP_525080.3 | 11 - 12 | 4627 | 66 | ||
NM_167164.4 | NP_727281.2 | 10 - 11 | 4546 | 66 | ||
GenBank mRNA | BT031006.1 | - | 9 - 10 | 3994 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |