Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rho-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080345.3 | NP_525084.1 | 3 - 4 | 1193 | 68 | |
NM_167290.3 | NP_727538.1 | 3 - 4 | 1193 | 68 | ||
GenBank mRNA | AF318285.1 | - | 3 - 4 | 1167 | 68 | |
AY061605.1 | - | 4 - 5 | 1497 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |