Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tango10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272532.2 | NP_001259461.1 | 4 - 5 | 1879 | 74 | |
NM_132515.3 | NP_572743.1 | 3 - 4 | 1691 | 74 | ||
NM_167302.3 | NP_727555.1 | 3 - 4 | 1930 | 74 | ||
GenBank mRNA | AY069304.1 | - | 3 - 4 | 1687 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |