Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PlexB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258471.2 | NP_001245400.1 | 2 - 3 | 5629 | 152 | |
NM_079877.4 | NP_524616.2 | 2 - 3 | 5629 | 152 | ||
GenBank mRNA | BT010009.1 | - | 2 - 3 | 5628 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |