Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eIF4G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103382.2 | NP_001096852.1 | 8 - 9 | 2217 | 108 | |
NM_001144385.2 | NP_001137857.1 | 7 - 8 | 1688 | 108 | ||
NM_079901.4 | NP_524640.3 | 7 - 8 | 1458 | 108 | ||
GenBank mRNA | AF030155.1 | - | 7 - 8 | 1464 | 108 | |
BT053671.1 | - | 7 - 8 | 1452 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Drosophila melanogaster |