Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300047.1 | NP_001286976.1 | 5 - 6 | 934 | 79 | |
NM_079922.4 | NP_524661.1 | 4 - 5 | 716 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |