Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079934.3 | NP_524673.3 | 1 - 2 | 492 | 85 | |
GenBank mRNA | AY069692.1 | - | 1 - 2 | 425 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |