Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259622.1 | NP_001246551.1 | 12 - 13 | 13014 | 70 | |
NM_001274375.1 | NP_001261304.1 | 11 - 12 | 12969 | 70 | ||
NM_001274376.1 | NP_001261305.1 | 11 - 12 | 12969 | 70 | ||
NM_001274377.1 | NP_001261306.1 | 11 - 12 | 12969 | 70 | ||
NM_001274378.1 | NP_001261307.1 | 11 - 12 | 12969 | 70 | ||
NM_001274379.1 | NP_001261308.1 | 11 - 12 | 12969 | 70 | ||
NM_001274380.1 | NP_001261309.1 | 11 - 12 | 12969 | 70 | ||
NM_001274381.1 | NP_001261310.1 | 11 - 12 | 12969 | 70 | ||
NM_001274383.1 | NP_001261312.1 | 11 - 12 | 12969 | 70 | ||
NM_001274384.1 | NP_001261313.1 | 11 - 12 | 12969 | 70 | ||
NM_079937.4 | NP_524676.2 | 11 - 12 | 12969 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |