Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103866.2 | NP_001097336.1 | 12 - 13 | 4149 | 86 | |
NM_001299556.1 | NP_001286485.1 | 11 - 12 | 4079 | 86 | ||
NM_001299557.1 | NP_001286486.1 | 12 - 13 | 6029 | 86 | ||
NM_137272.2 | NP_611116.2 | 12 - 13 | 4356 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |