Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032275.2 | NP_001027446.2 | 3 - 4 | 1762 | 68 | |
NM_001103925.3 | NP_001097395.1 | 3 - 4 | 1129 | 68 | ||
NM_001103926.4 | NP_001097396.3 | 4 - 5 | 1882 | 68 | ||
NM_001299741.1 | NP_001286670.1 | 3 - 4 | 1129 | 68 | ||
GenBank mRNA | KJ826443.1 | - | 2 - 3 | 367 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |