Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mamo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258733.1 | NP_001245662.1 | 2 | 625 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |