Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipsnap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298387.1 | NP_001285316.1 | 4 - 5 | 873 | 70 | |
NM_132875.4 | NP_573103.1 | 5 - 6 | 857 | 70 | ||
NM_167483.3 | NP_727931.1 | 5 - 6 | 898 | 70 | ||
NM_167484.3 | NP_727932.1 | 6 - 7 | 779 | 70 | ||
GenBank mRNA | AY061048.1 | - | 5 - 6 | 790 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |