Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201743.3 | NP_001188672.1 | 10 - 11 | 3159 | 83 | |
NM_001201744.2 | NP_001188673.1 | 6 - 7 | 1223 | 83 | ||
NM_001272779.1 | NP_001259708.1 | 10 - 11 | 3354 | 83 | ||
NM_134475.4 | NP_608319.1 | 6 - 7 | 1223 | 83 | ||
NM_167665.2 | NP_728267.1 | 10 - 11 | 2809 | 83 | ||
NM_167666.3 | NP_728268.1 | 10 - 11 | 3159 | 83 | ||
NM_167667.3 | NP_728269.1 | 5 - 6 | 1362 | 83 | ||
GenBank mRNA | AB277547.1 | - | 10 - 11 | 3138 | 83 | |
AB277548.1 | - | 6 - 7 | 1149 | 83 | ||
AY069469.1 | - | 6 - 7 | 1192 | 83 | ||
BT099719.1 | - | 10 - 11 | 3098 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |