Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201743.3 | NP_001188672.1 | 8 - 9 | 2861 | 80 | |
NM_001201744.2 | NP_001188673.1 | 4 - 5 | 925 | 80 | ||
NM_001272779.1 | NP_001259708.1 | 8 - 9 | 3056 | 80 | ||
NM_134475.4 | NP_608319.1 | 4 - 5 | 925 | 80 | ||
NM_167665.2 | NP_728267.1 | 8 - 9 | 2511 | 80 | ||
NM_167666.3 | NP_728268.1 | 8 - 9 | 2861 | 80 | ||
NM_167667.3 | NP_728269.1 | 3 - 4 | 1064 | 80 | ||
GenBank mRNA | AB277547.1 | - | 8 - 9 | 2840 | 80 | |
AB277548.1 | - | 4 - 5 | 851 | 80 | ||
AY069469.1 | - | 4 - 5 | 894 | 80 | ||
BT099719.1 | - | 8 - 9 | 2800 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |