Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: thoc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138167.1 | NP_612011.1 | 2 - 3 | 183 | 59 | |
NM_167804.2 | NP_728489.2 | 2 - 3 | 186 | 59 | ||
GenBank mRNA | AJ620302.1 | - | 2 - 3 | 183 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |