Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ns3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297850.1 | NP_001284779.1 | 3 - 4 | 1702 | 77 | |
NM_130559.3 | NP_569915.1 | 3 - 4 | 1702 | 77 | ||
GenBank mRNA | AY047556.1 | - | 3 - 4 | 1671 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |