Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fd3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001370027.1 | NP_001356931.1 | 3 - 4 | 1705 | 98 | |
NM_166991.3 | NP_726889.3 | 3 - 4 | 1705 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |