Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klp59C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137915.4 | NP_611759.1 | 1 | 1990 | 83 | |
GenBank mRNA | BT032681.1 | - | 1 | 1955 | 83 | |
DQ151904.1 | - | 1 | 1944 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |