Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: uri | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138089.4 | NP_611933.1 | 2 - 3 | 353 | 72 | |
GenBank mRNA | BT016108.1 | - | 2 - 3 | 325 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |