Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143682.4 | NP_651939.4 | 1 - 2 | 148 | 84 | |
GenBank mRNA | BT011201.1 | - | 1 - 2 | 150 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |