Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mei-P26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272447.2 | NP_001259376.1 | 6 - 7 | 3981 | 90 | |
NM_001272448.2 | NP_001259377.1 | 7 - 8 | 4442 | 90 | ||
NM_001272449.1 | NP_001259378.1 | 7 - 8 | 4419 | 90 | ||
NM_001298110.1 | NP_001285039.1 | 6 - 7 | 4017 | 90 | ||
NM_001369966.1 | NP_001356936.1 | 7 - 8 | 4283 | 90 | ||
NM_143765.4 | NP_652022.1 | 7 - 8 | 4283 | 90 | ||
GenBank mRNA | BT025212.1 | - | 7 - 8 | 4286 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |