Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144438.2 | NP_001137910.2 | 1 - 2 | 388 | 74 | |
NM_143787.3 | NP_652044.1 | 1 - 2 | 388 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |