Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274330.1 | NP_001261259.1 | 20 - 21 | 4316 | 60 | |
NM_001274331.1 | NP_001261260.1 | 20 - 21 | 4292 | 60 | ||
NM_139361.2 | NP_647618.1 | 19 - 20 | 4196 | 60 | ||
NM_167888.2 | NP_728620.1 | 19 - 20 | 4199 | 60 | ||
NM_167889.2 | NP_728621.1 | 21 - 22 | 4415 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |