Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258638.2 | NP_001245567.1 | 1 - 2 | 632 | 63 | |
NM_132176.4 | NP_572404.1 | 1 - 2 | 419 | 63 | ||
GenBank mRNA | AY060985.1 | - | 1 - 2 | 383 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |