Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258644.2 | NP_001245573.1 | 6 - 7 | 1507 | 58 | |
NM_132191.3 | NP_572419.1 | 4 - 5 | 1324 | 58 | ||
GenBank mRNA | AY051543.1 | - | 4 - 5 | 1291 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |