Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201629.2 | NP_001188558.1 | 7 - 8 | 2273 | 66 | |
NM_001201630.2 | NP_001188559.1 | 6 - 7 | 2252 | 66 | ||
NM_001201631.2 | NP_001188560.1 | 7 - 8 | 2053 | 66 | ||
NM_132230.4 | NP_572458.3 | 6 - 7 | 2252 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |