Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CG3776 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299939.1 | NP_001286868.1 | 3 - 4 | 750 | 76 | |
NM_138145.3 | NP_611989.1 | 2 - 3 | 683 | 76 | ||
GenBank mRNA | AY070543.1 | - | 2 - 3 | 667 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |