Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259601.2 | NP_001246530.1 | 14 - 15 | 5346 | 82 | |
NM_001274262.1 | NP_001261191.1 | 14 - 15 | 5349 | 82 | ||
NM_001274263.1 | NP_001261192.1 | 14 - 15 | 5397 | 82 | ||
NM_001274264.1 | NP_001261193.1 | 14 - 15 | 5382 | 82 | ||
NM_138175.4 | NP_612019.2 | 14 - 15 | 5400 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |