Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274263.1 | NP_001261192.1 | 8 - 9 | 3569 | 82 | |
NM_001274264.1 | NP_001261193.1 | 8 - 9 | 3572 | 82 | ||
NM_138175.4 | NP_612019.2 | 8 - 9 | 3572 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |