Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoGEF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144390.2 | NP_001137862.1 | 2 - 3 | 397 | 68 | |
NM_001274273.1 | NP_001261202.1 | 11 - 12 | 8163 | 68 | ||
NM_138180.4 | NP_612024.4 | 11 - 12 | 8166 | 68 | ||
NM_138182.2 | NP_612026.2 | 2 - 3 | 553 | 68 | ||
NM_138183.3 | NP_612027.2 | 2 - 3 | 397 | 68 | ||
GenBank mRNA | BT044233.1 | - | 2 - 3 | 377 | 68 | |
BT099707.1 | - | 2 - 3 | 553 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |