Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138199.3 | NP_612043.1 | 2 - 3 | 314 | 59 | |
GenBank mRNA | U65492.1 | - | 2 - 3 | 353 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |