Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mwh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274314.1 | NP_001261243.1 | 4 - 5 | 1450 | 65 | |
NM_001274315.1 | NP_001261244.1 | 4 - 5 | 1453 | 65 | ||
NM_138238.3 | NP_612082.1 | 2 - 3 | 941 | 65 | ||
GenBank mRNA | AM231270.1 | - | 2 - 3 | 926 | 65 | |
BT023932.1 | - | 2 - 3 | 938 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |