Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oseg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274349.1 | NP_001261278.1 | 8 - 9 | 3603 | 84 | |
NM_139396.4 | NP_647653.1 | 8 - 9 | 3603 | 84 | ||
GenBank mRNA | AY061373.1 | - | 8 - 9 | 3462 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |