Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cht7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139511.4 | NP_647768.3 | 6 - 7 | 2922 | 74 | |
GenBank mRNA | AY051988.1 | - | 6 - 7 | 2895 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |