Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274747.1 | NP_001261676.1 | 3 - 4 | 587 | 63 | |
NM_168397.2 | NP_729591.1 | 3 - 4 | 587 | 63 | ||
NM_168398.2 | NP_729592.1 | 3 - 4 | 510 | 63 | ||
NM_168399.2 | NP_729593.1 | 3 - 4 | 670 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |