Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vha16-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202157.1 | NP_001189086.1 | 2 | 309 | 84 | |
NM_168458.2 | NP_729706.1 | 1 | 263 | 84 | ||
GenBank mRNA | KX531470.1 | - | 1 | 241 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |