Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bru3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202161.2 | NP_001189090.1 | 2 - 3 | 818 | 96 | |
NM_168557.3 | NP_729923.1 | 2 - 3 | 818 | 96 | ||
GenBank mRNA | AY060398.1 | - | 2 - 3 | 818 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |