Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: futsch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103394.3 | NP_001096864.1 | 6 - 7 | 15361 | 64 | |
NM_001169161.3 | NP_001162632.2 | 7 - 8 | 15997 | 64 | ||
NM_001258532.1 | NP_001245461.1 | 7 - 8 | 16797 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |