Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: msk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080041.3 | NP_524780.1 | 4 - 5 | 1257 | 71 | |
GenBank mRNA | AF132299.1 | - | 4 - 5 | 1166 | 71 | |
BT053682.1 | - | 4 - 5 | 1258 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |