Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kto | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259906.1 | NP_001246835.1 | 2 - 3 | 347 | 47 | |
NM_080047.2 | NP_524786.1 | 7 - 8 | 6364 | 47 | ||
GenBank mRNA | AF324426.1 | - | 7 - 8 | 6364 | 47 | |
BT150248.1 | - | 2 - 3 | 334 | 47 | ||
BT150292.1 | - | 2 - 3 | 334 | 47 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |