Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042804.3 | NP_001036269.2 | 6 - 7 | 8740 | 58 | |
NM_001272506.1 | NP_001259435.1 | 3 - 4 | 3349 | 58 | ||
NM_001298174.1 | NP_001285103.1 | 5 - 6 | 8564 | 58 | ||
NM_132441.3 | NP_572669.2 | 5 - 6 | 8467 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |