Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042804.3 | NP_001036269.2 | 7 - 8 | 9891 | 76 | |
NM_001272506.1 | NP_001259435.1 | 4 - 5 | 4500 | 76 | ||
NM_001298174.1 | NP_001285103.1 | 6 - 7 | 9715 | 76 | ||
NM_132441.3 | NP_572669.2 | 6 - 7 | 9618 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |